Learn More
OTX2 Monoclonal Antibody (1H12G8B2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515855
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
MA5-15855 targets OTX2 in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15855 immunogen is purified recombinant fragment of human OTX2 expressed in E. Coli. . MA5-15855 detects OTX2 which has a predicted molecular weight of approximately 32kDa.
OTX2 Monoclonal antibody specifically detects OTX2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot.Spezifikation
OTX2 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P32243, P80206, Q64201 | |
OTX2 | |
Purified recombinant fragment of human OTX2 expressed in E. Coli. | |
RUO | |
18424, 305858, 5015 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
1H12G8B2 | |
Unconjugated | |
OTX2 | |
CPHD6; E130306E05Rik; homeobox Otx2; homeobox protein OTX2; MCOPS5; orthodenticle homeobox 2; orthodenticle homolog 2; OTX; OTX2 | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |