Learn More
OVOL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541480
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: QKYAYKERRA KLYVCEECGC TSESQEGHVL HLKEHHPDSP LLRKTSKKVA Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%
OVOL1 Polyclonal antibody specifically detects OVOL1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
OVOL1 | |
Polyclonal | |
Unconjugated | |
OVOL1 | |
BB147136; CD254; hOvo1; hRANKL2; Ly109l; movo1; mOvo1a; ODF; OPG; OPG ligand; OPGL; OPTB2; osteoclast differentiation factor; Osteoprotegerin ligand; OVO homolog-like 1; OVO homolog-like 1 (Drosophila); ovo like transcriptional repressor 1; ovo like zinc finger 1; Ovo1; Ovol1; OVO-like 1 binding protein; ovo-like 1(Drosophila); ovo-like zinc finger 1; Putative transcription factor Ovo-like 1; RANKL; receptor activator of NF-kappaB ligand; receptor activator of nuclear factor kappa B ligand; receptor activator of nuclear factor kappa-B ligand; sOdf; TNF-related activation-induced cytokine; TNFSF11; TNLG6B; Trance; transcription factor Ovo-like 2; tumor necrosis factor (ligand) superfamily member 11; tumor necrosis factor (ligand) superfamily, member 11; tumor necrosis factor ligand 6B; tumor necrosis factor ligand superfamily member 11; Tumor necrosis factor ligand superfamily member 11, membrane form; Tumor necrosis factor ligand superfamily member 11, soluble form; tumor necrosis fact | |
Rabbit | |
Protein A | |
RUO | |
5017 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O14753 | |
OVOL1 | |
Synthetic peptide directed towards the C-terminal of human OVOL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |