missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OVOL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541619
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: PKVFLVKRRS LGVSVRSWDE LPDEKRADTY IPVGLGRLLH DPPEDCRSDG Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 93%
OVOL2 Polyclonal antibody specifically detects OVOL2 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
OVOL2 | |
Polyclonal | |
Unconjugated | |
OVOL2 | |
1700108N11Rik; 1810007D21Rik; EUROIMAGE566589; hOvo2; MOVO; M-OVO; movo2; M-OVO-A; M-OVO-B; MOVO-B zinc finger protein; MOVO-C zinc finger protein; OVO homolog-like 2; ovo like zinc finger 2; Ovo2; OVOL2; ovo-like 2; ovo-like 2 (Drosophila); ovo-like zinc finger 2; PPCD1; transcription factor Ovo-like 2; Zfp339; zinc finger OVO2; Zinc finger protein 339; zinc finger protein mOVO; Znf339 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
58495 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BRP0 | |
OVOL2 | |
Synthetic peptide directed towards the N-terminal of human OVOL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |