missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXER1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534062
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey (100%).
OXER1 Polyclonal antibody specifically detects OXER1 in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
OXER1 | |
Polyclonal | |
Unconjugated | |
OXER1 | |
5-oxo-ETE acid G-protein-coupled receptor 1; 5-oxo-ETE G-protein coupled receptor; GPCR; GPR170; G-protein coupled receptor 170; G-protein coupled receptor R527; G-protein coupled receptor TG1019; HEOAD54; OXE receptor; OXER1; oxoeicosanoid (OXE) receptor 1; oxoeicosanoid receptor 1; TG1019 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
165140 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8TDS5 | |
OXER1 | |
Synthetic 19 amino acid peptide from C-terminus of human OXER1. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |