Learn More
OXNAD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532011
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: U87-MG. Predicted reactivity: Mouse (83%), Rat (85%), Rhesus Monkey (96%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
OXNAD1 Polyclonal antibody specifically detects OXNAD1 in Human samples. It is validated for Western Blot.Spezifikation
OXNAD1 | |
Polyclonal | |
Unconjugated | |
OXNAD1 | |
2410002F01Rik; AW492292; BC019806; oxidoreductase NAD binding domain containing 1; oxidoreductase NAD-binding domain containing 1; oxidoreductase NAD-binding domain-containing protein 1; Oxnad1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92106 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96HP4 | |
OXNAD1 | |
Recombinant fragment corresponding to a region within amino acids 2 and 312 of Human OXNAD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |