Learn More
P-Selectin/CD62P Rat anti-Mouse, Clone: 127933, R&D Systems™
Rat Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB737
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411756
  • 510.09 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 29-10-2019
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
P-Selectin/CD62P Monoclonal antibody MAB737 Specifically detects P-Selectin/CD62P in Mouse samples. It is validated for ELISA Capture, Western Blotting
Spezifikation
Spezifikation
P-Selectin/CD62P
127933
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q01102
CD62P, CD62P antigen, FLJ45155, GMP140, GRMP, PADGEM, PADGEMantigen CD62), PSEL, selectin P (granule membrane protein 140kDa, antigen CD62), SELP
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse P-Selectin/CD62P in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) L-Selectin, rmE-Selectin, or recombinant human P-Selectin is observed.
Mouse
ELISA, Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SELP
Mouse myeloma cell line NS0-derived recombinant mouse P-Selectin/CD62P Trp42-Ala709 Accession # Q01102
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
6403
Reconstitute at 0.5 mg/mL in sterile PBS.