Learn More
p14ARF Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA131212
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-31212 detects p14ARF from rat, human, mouse samples. PA1-31212 has been successfully used in Western blot applications. Predicted molecular weight: 17 kDa. The PA1-31212 immunogen is: Synthetic peptide: GRCVGPSLQLRGQEWRCS (Human) (N terminal). Store at 4°C short term. For extended storage aliquot and store at -20°C or below. Avoid freeze-thaw cycles.
The INK4a locus on chromosome 9p21 is frequently affected in human tumors. It encodes two structurally distinct tumor suppressor proteins, p16INK4a and the alternative reading frame protein, ARF (p14ARF in human and P19ARF in mouse). These two proteins interact with the upstream proteins: retinoblastoma protein and p53 tumor suppressors, respectively. Each of these proteins has a role in the senescence of primary cells, activates pathways for cell cycle control and tumor suppression. The p14ARF (or P19ARF in mouse) proteins can induce both G1 and G2 phase arrest in a manner that depends on functional p53. The mouse and human ARF proteins have different functions in tumor suppression; this distinction may contribute to the different levels of tumor proneness of these species.Spezifikation
p14ARF | |
Polyclonal | |
Unconjugated | |
P42771, P51480, Q8QZZ9 | |
CDKN2A | |
synthetic peptide GRCVGPSLQLRGQEWRCS (Human) (N terminal). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
0.65 mg/mL | |
CDKN2A | |
ARF, CDK4I, CDKN2, CMM2, INK4, INK4a, MLM, MTS1, TP16, p14, p14ARF, p16, p16INK4, p16INK4a, p19, p19Arf | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1029, 12578, 25163 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |