missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p27 Kip1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA517946
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology.
p27 Kip1 Polyclonal antibody specifically detects p27 Kip1 in Human samples. It is validated for Immunoprecipitation, Western Blot.Spezifikation
p27 Kip1 | |
Polyclonal | |
Unconjugated | |
Cdkn1b | |
AA408329; AI843786; Cdki1b; CDKN1B; CDKN4; CDN1B; cyclin dependent kinase inhibitor 1B; cyclin-dependent kinase inhibitor 1B; Cyclin-dependent kinase inhibitor 1B (p27 Kip1); Cyclin-dependent kinase inhibitor 1B (p27, Kip1); cyclin-dependent kinase inhibitor p27; Cyclin-dependent kinase inhibitor p27 Kip1; cyclin-dependent kinase inhibitor p27/Kip1; KIP1; MEN1B; MEN4; p27; P27 CDKN1B; p27(KIP1); p27Kip1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
1027 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P46527 | |
Cdkn1b | |
Synthetic peptide sequence (EQTPKKPGLRRRQT) corresponding to the C-terminus amino acids of CDKN1B | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |