missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532201
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: mouse brain. Predicted reactivity: Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
P2X7 Polyclonal antibody specifically detects P2X7 in Mouse, Rat samples. It is validated for Western Blot.Spezifikation
P2X7 | |
Polyclonal | |
Unconjugated | |
P2RX7 | |
AI467586; ATP receptor; MGC20089; P2rx7; P2X purinoceptor 7; p2X receptor 7; P2X(7); P2X7; P2X7 purinoceptor; P2X7 receptor; P2X7R; P2Z; P2Z receptor; Purinergic receptor; purinergic receptor P2X 7; purinergic receptor P2X, ligand gated ion channel, 7; purinergic receptor P2X, ligand-gated ion channel, 7; purinergic receptor P2X7; purinergic receptor P2X7 variant A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18439, 29665 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q64663, Q9Z1M0 | |
P2RX7 | |
Synthetic peptide corresponding to a region within amino acids 526 and 595 of Rat P2rx7 | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |