missing translation for 'onlineSavingsMsg'
Learn More

P2X2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA531318

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13415837

  • 470.00 € / 100 Mikroliter
Verfügbar ab: 08-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: A549, HepG2, mouse brain. Predicted reactivity: Mouse (81%), Rat (80%), Bovine (86%), Guinea pig (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

P2X2 Polyclonal antibody specifically detects P2X2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

P2X2
Polyclonal
Unconjugated
P2RX2
ATP receptor; DFNA41; MGC129601; P2RX2; P2X purinoceptor 2; P2X Receptor, subunit 2; P2X2; P2X2 purinoceptor; P2X2 receptor splice variant P2X2-1; P2X2 receptor splice variant P2X2-2; P2X2 receptor splice variant P2X2-3; P2X2a; Purinergic receptor; purinergic receptor P2X 2; purinergic receptor P2X, ligand gated ion channel, 2; purinergic receptor P2X, ligand-gated ion channel, 2; purinergic receptor P2X2
Rabbit
Antigen affinity chromatography
RUO
22953, 231602
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q8K3P1, Q9UBL9
P2RX2
Recombinant fragment corresponding to a region within amino acids 8 and 302 of Human P2X2
100 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote