missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534075
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Hamster (100%) Gibbon, Mouse, Rat, Panda, Bat, Bovine, Rabbit, Horse (94%).
P2Y10 Polyclonal antibody specifically detects P2Y10 in Hamster, Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
P2Y10 | |
Polyclonal | |
Unconjugated | |
P2RY10 | |
5830408N17Rik; G-protein coupled purinergic receptor P2Y10; LYPSR2; P2RY10; p2Y purinoceptor 10; P2Y10; P2Y-like receptor; purinergic receptor P2Y, G-protein coupled 10; purinergic receptor P2Y, G-protein coupled, 10; purinergic receptor P2Y10; putative P2Y purinoceptor 10; RGD1561912; RP3-333E23.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27334 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O00398 | |
P2RY10 | |
Synthetic 18 amino acid peptide from C-terminus of human P2RY10 / P2Y10. | |
50 μg | |
Primary | |
Hamster, Human, Non-human Primate | |
Antibody | |
IgG |