missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA120615
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Positive control: Dendritic cells
P2Y11 Polyclonal antibody specifically detects P2Y11 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
P2Y11 | |
Polyclonal | |
Unconjugated | |
P2RY11 | |
P2RY11; P2Y purinoceptor 11; P2Y11; purinergic receptor P2Y, G-protein coupled, 11; purinergic receptor P2Y11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5032 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q96G91 | |
P2RY11 | |
Synthetic 16 amino acid peptide from 3rd extracellular domain of human P2Y11 | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |