missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534091
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey (100%) Mouse, Rat, Hamster, Panda, Bovine (93%) Rabbit, Pig, Opossum (86%).
P2Y4 Polyclonal antibody specifically detects P2Y4 in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
P2Y4 | |
Polyclonal | |
Unconjugated | |
P2RY4 | |
NRU; P2P; P2RY4; P2Y ATP receptor 4; P2Y purinoceptor 4; P2Y4; P2Y4 receptor; P2Y4R; P2YR4; purinergic receptor P2Y G-protein coupled 4; purinergic receptor P2Y, G-protein coupled 4; purinergic receptor P2Y, G-protein coupled, 4; pyrimidinergic receptor P2Y, G-protein coupled, 4; pyrimidinergic receptor P2Y4; RP13-26D14.5; UNR; Uridine nucleotide receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5030 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P51582 | |
P2RY4 | |
Synthetic 14 amino acid peptide from 1st cytoplasmic domain of human P2RY4 / P2Y4. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |