Learn More
CBP Monoclonal Antibody (NM11), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA513634
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-13634 targets p300/CBP (CREB-Binding Protein) in ICC/IF and IHC (P) applications and shows reactivity with Human, Mink, mouse, Non-human primate, and Rat samples. The MA5-13634 immunogen is e1A affinity purified p300 protein.
CBP Monoclonal antibody specifically detects CBP in Human, Mink, Mouse, Non-human Primate, Rat samples. It is validated for ChIP Assay, Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
CBP | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P45481, Q6JHU9, Q92793 | |
CREBBP | |
E1A affinity purified p300 protein | |
500 μL | |
Primary | |
Human, Mink, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG2b κ |
ChIP Assay, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
NM11 | |
Unconjugated | |
CREBBP | |
AW558298; CBP; CBP/p300; CREB binding protein; CREB-binding protein; CREBBP; KAT3A; p300/CBP; RSTS; RTS | |
Mouse | |
Protein A | |
RUO | |
12914, 1387, 54244 | |
4° C | |
Liquid |