missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12 p35 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA523226
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
IL-12 p35 Polyclonal antibody specifically detects IL-12 p35 in Human samples. It is validated for Western Blot.Spezifikation
IL-12 p35 | |
Polyclonal | |
Unconjugated | |
IL12A | |
CLMF; CLMF p35; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; il 12; IL-12 subunit p35; IL-12, subunit p35; Il12a; IL-12A; IL12p35; IL-12p35; IL35 subunit; ILN; Interleukin; Interleukin 12; interleukin 12 p35 subunit; interleukin 12, p35; interleukin 12a; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); Interleukin12; interleukin-12 alpha chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit; interleukin-12 subunit alpha; Ll12a; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NKSF1; P35 | |
Rabbit | |
Protein A | |
RUO | |
3592 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
P29459 | |
IL12A | |
A portion of amino acids 200-219 of IL-12A | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |