Learn More
ERK1/ERK2 Mouse anti-Human, Mouse, Rat, Clone: 3A1, Invitrogen™
Mouse Monoclonal Antibody
Marke:  Invitrogen LFMA0178
Weitere Details : Netto-Gewicht : 0.99000kg
Artikelnummer. 10889230
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Beschreibung
Beschreibung
A suggested positive control for this product is HepG2 cells.
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act in a signaling cascade that regulates various cellular processes such as proliferation, differentiation, and cell cycle progression in response to a variety of extracellular signals. This kinase is activated by upstream kinases, resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Alternatively spliced transcript variants encoding different protein isoforms have been described.
Spezifikation
Spezifikation
ERK1/ERK2
Monoclonal
1 mg/mL
MAPK1
Mitogen-activated protein kinase 1, MAP kinase 1, MAPK 1, ERT1, Extracellular signal-regulated kinase 2, ERK-2, MAP kinase isoform p42, p42-MAPK, Mitogen-activated protein kinase 2, MAP kinase 2, MAPK 2 , MAPK1, ERK2, PRKM1, PRKM2, Mitogen-activated protein kinase 3, MAP kinase 3, MAPK 3, ERT2, Extracellular signal-regulated kinase 1, ERK-1, Insulin-stimulated MAP2 kinase, MAP kinase isoform p44, p44-MAPK, Microtubule-associated protein 2 kinase, p44-ERK1, MAPK3, ERK1, PRKM3, p44, p42
Mouse
Protein G
RUO
116590, 26413, 26417, 50689, 5594, 5595
-20° C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
3A1
Unconjugated
P27361, P28482, P63085, Q63844, P21708, P63086
MAPK1, MAPK3
His-tagged recombit human ERK1 protein purified from E.coli
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG1