missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p47phox Monoclonal Antibody (G.207.2), Invitrogen™
Rabbit Monoclonal Antibody
Marke: Invitrogen MA514778
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
p47phox Monoclonal antibody specifically detects p47phox in Human samples. It is validated for Western Blot.Spezifikation
p47phox | |
Monoclonal | |
35 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P14598 | |
Ncf1 | |
Synthetic peptide corresponding to residues around Asp217 of human p47phox | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
G.207.2 | |
Unconjugated | |
Ncf1 | |
47 kDa autosomal chronic granulomatous disease protein; 47 kDa neutrophil oxidase factor; NADPH oxidase organizer 2; NADPH oxidase subunit (47 kD); NADPH oxidase subunit (47kDa); Ncf1; NCF-1; NCF1A; NCF-47K; neutrop; neutrophil cytosol factor 1; neutrophil cytosolic factor 1; neutrophil cytosolic factor 1, (chronic granulomatous disease, autosomal 1); neutrophil NADPH oxidase factor 1; Nox organizer 2; NOXO2; nox-organizing protein 2; P47 phox; p47<phox>; p47phox; p47-phox; phagocyte oxidase 47 kDa; SH3 and PX domain-containing protein 1A; SH3PXD1A | |
Rabbit | |
Affinity chromatography | |
RUO | |
653361 | |
-20°C | |
Liquid |