Learn More
P4HA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531246
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: PC-3. Predicted reactivity: Mouse (97%), Rat (95%), Pig (99%), Chicken (96%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
P4HA1 Polyclonal antibody specifically detects P4HA1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
P4HA1 | |
Polyclonal | |
Unconjugated | |
P4HA1 | |
4-PH alpha-1; AL022634; collagen prolyl 4-hydroxylase alpha(I); C-P4Halpha(I); P4ha; P4ha1; PHalphaI; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha 1 polypeptide; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide I; procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-1; prolyl 4-hydroxylase alpha subunit; prolyl 4-hydroxylase subunit alpha 1; prolyl 4-hydroxylase subunit alpha-1; prolyl 4-hydroxylase, alpha polypeptide I | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5033 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P13674 | |
P4HA1 | |
Recombinant fragment corresponding to a region within amino acids 318 and 534 of Human P4HA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |