Learn More
PDIA6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA3008
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA3-008 detects P5 protein in human samples. PA3-008 has successfully been used in Western blot procedures. By Western blot, this antibody detects an ∽50 kDa protein representing P5 in human fibrosarcoma HT 1080 cell lysate. The PA3-008 immunogen is a synthetic peptide corresponding to residues D(476) I D L S D V E L D D L G K D E L(492) of human P5. This peptide (Cat. # PEP-224) is available for use in neutralization and control experiments.
PDIA6 Polyclonal antibody specifically detects PDIA6 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
PDIA6 | |
Polyclonal | |
Unconjugated | |
PDIA6 | |
1700015E05Rik; AL023058; C77895; CaBP1; CaBP5; Calcium-binding protein 1; endoplasmic reticulum protein 5; ER protein 5; ERP5; P5; PDIA6; pdi-p5; protein disulfide isomerase A6; protein disulfide isomerase associated 6; protein disulfide isomerase family A member 6; protein disulfide isomerase family A, member 6; protein disulfide isomerase P5; protein disulfide isomerase-associated 6; protein disulfide isomerase-related protein; protein disulfide-isomerase A6; thioredoxin domain containing 7; thioredoxin domain containing 7 (protein disulfide isomerase); thioredoxin domain-containing protein 7; TXNDC7 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Conc. Not Determined | |
whole serum with 0.01% sodium azide | |
Q15084 | |
PDIA6 | |
Synthetic peptide corresponding to residues D(476) I D L S D V E L D D L G K D E L(492) of human P5. | |
RUO | |
10130 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |