missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 Monoclonal Antibody (184727), APC, Invitrogen™
Mouse Monoclonal Antibody
Marke: Thermo Scientific MA523670
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
p53 Monoclonal antibody specifically detects p53 in Human samples. It is validated for Flow Cytometry.Spezifikation
p53 | |
Monoclonal | |
APC | |
Tp53 | |
Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3 | |
Mouse | |
Protein A/G | |
RUO | |
7157 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
184727 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
P04637 | |
Tp53 | |
E. coli-derived recombinant human p53 Asp7-Asp393 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b |