Learn More
p53 Monoclonal Antibody (DO-7), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA512554
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-12554 targets p53 in ICC/IF, FACS, IHC (P), IP, and WB applications and shows reactivity with Bovine, Human, and Non-human primate samples. The MA5-12554 immunogen is recombinant human wild-type p53 protein expressed in E. coli.
p53 Monoclonal antibody specifically detects p53 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
p53 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P04637 | |
Tp53 | |
Recombinant human wild-type p53 protein expressed in E. coli | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
DO-7 | |
Biotin | |
Tp53 | |
Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3 | |
Mouse | |
Protein A | |
RUO | |
7157 | |
4° C | |
Liquid |