Learn More
p53 Monoclonal Antibody (4A8), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515510
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15510 targets p53 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15510 immunogen is purified recombinant fragment of human p53 expressed in E. Coli. MA5-15510 detects p53 which has a predicted molecular weight of approximately 43.7kDa.
p53 Monoclonal antibody specifically detects p53 in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
p53 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P04637 | |
Tp53 | |
Purified recombinant fragment of human p53 expressed in E. Coli. | |
RUO | |
7157 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
4A8 | |
Unconjugated | |
Tp53 | |
Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |