Learn More
p53 Mouse, Unlabeled, Clone: 80, BD
Mouse Monoclonal Antibody
499.27 €
Spezifikation
Antigen | p53 |
---|---|
Klon | 80 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15895309
|
BD Biosciences
610184 |
150 μg |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
The p53 protein is critical to regulation of normal cell growth and is a suppressor of tumor cell proliferation. Inactivation of p53 by a number of mechanisms, such as missense mutations or interaction with oncogenic viral or cellular proteins, can result in tumor progression. Mutations and/or allelic loss of the p53 gene is associated with a wide variety of human tumors. Known to have a role in transcriptional regulation, p53 suppresses various promoters containing TATA elements in an apparently sequence-independent fashion. p53 also binds to DNA in a sequence-specific manner via recognition of a 20 bp consensus-binding site. This interaction stimulates the expression of genes downstream of the p53 binding site. A number of genes that contain p53-binding sites have been identified, including MDM2, GADD45, and muscle creatine kinase. It is thought that MDM2 is a feedback regulator of p53. In addition, a p53-inducible gene, Cip1, has been identified and shown to suppress tumor cell growth in culture.
Spezifikation
p53 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2b κ | |
Affinity Purified |
80 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human | |
IgG2a, κ | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.