missing translation for 'onlineSavingsMsg'
Learn More

p53 Monoclonal Antibody (Y5), Invitrogen™

Rabbit Monoclonal Antibody

Marke:  Invitrogen MA514467

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11529201

  • 918.00 € / 1 Milliliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

MA5-14467 targets p53 in IHC (P) applications and shows reactivity with Human samples. The MA5-14467 immunogen is a synthetic peptide derived from N- terminal residues of human p53.

p53 Monoclonal antibody specifically detects p53 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

p53
Monoclonal
Conc. Not Determined
tissue culture supernatant with 15mM sodium azide
P04637
Tp53
A synthetic peptide derived from N- terminal residues of human p53
RUO
7157
4° C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Y5
Unconjugated
Tp53
Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3
Rabbit
1 mL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote