missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFIT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA3848
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA3-848 detects ISG56 (p56) from human and mouse samples. PA3-848 has been successfully used in Western blot and ICC/IF applications to detect a 39-58 kDa band.
IFIT1 Polyclonal antibody specifically detects IFIT1 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
IFIT1 | |
Polyclonal | |
Unconjugated | |
IFIT1 | |
C56; G10P1; Garg16; GARG-16; glucocorticoid-attenuated response gene 16 product; glucocorticoid-attenuated response gene 16 protein; Ifi56; IFI-56; IFI-56K; IFIT1; IFIT-1; IFNAI1; interferon induced protein with tetratricopeptide repeats 1; interferon, alpha-inducible protein (MW 56kD); interferon-induced 56 kDa protein; Interferon-induced protein with tetratricopeptide repeats 1; interferon-induced protein with tetratricopeptide repeats 1-like protein; interferon-inducible mRNA 561; Isg56; P56; RNM561 | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Conc. Not Determined | |
whole serum with 0.05% sodium azide | |
P09914, Q64282 | |
IFIT1 | |
Bacterially expressed full-length human p56 protein | |
RUO | |
15957, 3434 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |