Learn More
p57 Kip2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532532
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Heat-mediated antigen retrieval is recommended prior to staining, using a 1mM EDTA buffer, pH 8.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is placenta or hydatid mole.
p57 Kip2 Polyclonal antibody specifically detects p57 Kip2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
p57 Kip2 | |
Polyclonal | |
Unconjugated | |
CDKN1C | |
AL024410; Beckwith Wiedemann syndrome (WBS); BWCR; BWS; CDKI; Cdkn1c; cyclin dependent kinase inhibitor 1C; Cyclin dependent kinase inhibitor 1C (CDKN1C); cyclin-dependent kinase inhibitor 1C; cyclin-dependent kinase inhibitor 1C (P57); cyclin-dependent kinase inhibitor 1C (p57, Kip2); cyclin-dependent kinase inhibitor p57; IMAGE; KIP2; p57; p57 Kip2; p57(kip2); p57KIP2; p57KIP2 A3sh; WBS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1028 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.021 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
P49918 | |
CDKN1C | |
Synthetic peptide derived from N-terminus of human p57/kip2 protein. | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |