missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATAD2B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA141333
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Suggested positive control: HeLa or RAW.
GATAD2B Polyclonal antibody specifically detects GATAD2B in Human, Mouse samples. It is validated for Western Blot.Spezifikation
GATAD2B | |
Polyclonal | |
Unconjugated | |
GATAD2B | |
AL118180; C430014D17Rik; GATA zinc finger domain containing 2B; GATA zinc finger domain-containing protein 2B; GATAD2B; KIAA1150; mKIAA1150; MRD18; p66/p68; P66beta; p68; RGD1308533; transcription repressor p66 beta component of the MeCP1 complex; transcription repressor p66 component of the MeCP1 complex; transcriptional repressor p66-beta | |
Rabbit | |
Protein G | |
RUO | |
229542, 57459 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
Q8VHR5, Q8WXI9 | |
GATAD2B | |
Synthetic peptide corresponding to residues 445-460 of mouse p66b. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |