missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p73 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA112695
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control for this product is COS or IM9 myeloma cells.
p73 Polyclonal antibody specifically detects p73 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.Spezifikation
p73 | |
Polyclonal | |
Unconjugated | |
TP73 | |
deltaNp73; OTTHUMP00000003479; OTTHUMP00000003480; p53-like transcription factor; p53-related protein; P73; RP23-254N4.10; TAp73; Tp73; transformation related protein 73; Trp73; Tumor protein p73 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
22062, 7161 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.08% sodium azide | |
O15350, Q9JJP2 | |
TP73, Trp73 | |
GST-fused recombinant protein corresponding to the C-terminal of the p73 protein. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |