missing translation for 'onlineSavingsMsg'
Learn More

NGFR Monoclonal Antibody (MC192), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen OSM00019A

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11514853

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

In IHC applications, this antibody is suitable for use in paraformaldehyde-fixed, alcohol-fixed and paraffin-embedded sections. Reconstitute in 100 μL of sterile water.

Low affinity receptor which can bind to NGF, BDNF, NT-3, and NT-4. Can mediate cell survival as well as cell death of neural cells. Homodimer; disulfide-linked. Interacts with p75NTR-associated cell death executor. Interacts with NGFRAP1/BEX3. Interacts with TRAF2, TRAF4, TRAF6, PTPN13 and RANBP9. Interacts through TRAF6 with SQSTM1 which bridges NGFR to NTRK1. Interacts with BEX1.
Spezifikation

Spezifikation

NGFR
Monoclonal
1 mg/mL
Ngfr
TNR16, low-affinity nerve growth factor receptor, NGF receptor, Gp80-LNGFR, p75 ICD, low affinity neurotrophin receptor p75NTR, Tnfrsf16, Ngfr, p75, L, p75, p-75
Mouse
Affinity chromatography
RUO
24596
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
MC192
Unconjugated
P07174
NGFR
Nerve growth factor receptor
100 μg
Primary
Rat
Antibody
IgG1
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote