Learn More
alpha Synuclein Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA118256
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-18256 detects alpha, beta, and gamma Synuclein from human and rat samples. Reconstitute with 100 μL of distilled water. After reconstitution, store aliquots at -20oC for a higher stability, and at 4oC with an appropriate antibacterial agent.
alpha Synuclein Polyclonal antibody specifically detects alpha Synuclein in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Spezifikation
alpha Synuclein | |
Polyclonal | |
Unconjugated | |
SNCA | |
Alpha synuclein; alphaSYN; alpha-synuclein; NACP; non A-beta component of AD amyloid; non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; PD1; Snca; Syn; synuclein alpha; synuclein alpha-140; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor) | |
Sheep | |
Affinity chromatography | |
RUO | |
20617, 29219, 6622 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability. | |
Lyophilized |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O55042, P37377, P37840 | |
SNCA | |
A synthetic peptide (AKEGVVAAAEKTKQGV) identical in human alpha, beta, and gamma synuclein conjugated to diphteria toxoid | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |