missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF-A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA511722
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PDGF-A Polyclonal antibody specifically detects PDGF-A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PDGF-A | |
Polyclonal | |
Unconjugated | |
PDGFA | |
PDGF A-chain; PDGF subunit A; PDGF1; PDGF-1; Pdgfa; PDGF-A; PDGFACP; platelet derived growth factor subunit A; platelet derived growth factor, alpha; platelet-derived growth factor A chain; platelet-derived growth factor A-chain; platelet-derived growth factor alpha chain; platelet-derived growth factor alpha polypeptide; Platelet-derived growth factor subunit A; Rpa1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
5154 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P04085 | |
PDGFA | |
KLH conjugated synthetic peptide between 60-89 amino acids from the N-terminal region of human PDGFA | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |