missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pyruvate dehydrogenase kinase, isozyme 3, Mouse, Clone: 3A1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PDK3.
Marke: Abnova H00005165-M02.100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: GDTNPVHPKHIGSIDPTCNVADVVKDAYETAKMLCEQYYLVAPELEVEEFNAKAPDKPIQVVYVPSHLFHMLFELFKNSMRATVELYEDRSpezifikation
pyruvate dehydrogenase kinase, isozyme 3 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC015948 | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
5165 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
ELISA, Western Blot | |
3A1 | |
Mouse monoclonal antibody raised against a partial recombinant PDK3. | |
PDK3 | |
PDK3 | |
PDK3 (AAH15948, 174 a.a. ∼ 263 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Kinases | |
Primary | |
Human | |
Antibody |