Learn More
PGD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521376
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, Molt-4, Raji, Mouse heart, Mouse lung. Predicted reactivity: Mouse (97%), Rat (95%), Zebrafish (86%), Xenopus laevis (85%), Chicken (90%), Sheep (94%), Chimpanzee (100%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PGD Polyclonal antibody specifically detects PGD in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PGD | |
Polyclonal | |
Unconjugated | |
PGD | |
0610042A05Rik; 6PGD; 6-phosphogluconate dehydrogenase, decarboxylating; 6-phosphogluconate dehydrogenase, decarboxylating-like; AU019875; C78335; LOC100360180; Pgd; PGDH; phosphogluconate dehydrogenase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
110208, 5226 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P52209, Q9DCD0 | |
PGD | |
Recombinant fragment corresponding to a region within amino acids 1 and 201 of PGD (Uniprot ID#P52209) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |