missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-beta Catenin (Thr41, Ser45) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517916
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
Phospho-beta Catenin (Thr41, Ser45) Polyclonal antibody specifically detects Phospho-beta Catenin (Thr41, Ser45) in Canine, Human, Mouse, Non-human Primate, Porcine samples. It is validated for Western Blot.Spezifikation
Phospho-beta Catenin (Thr41, Ser45) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
B6V8E6, P35222, Q02248 | |
CTNNB1 | |
Synthetic phosphopeptide corresponding to residues surrounding pThr41 and pSer45 of human β-catenin | |
100 μL | |
Primary | |
Canine, Human, Mouse, Non-human Primate, Porcine | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CTNNB1 | |
armadillo; armadillo homolog; Bcatenin; b-catenin; beta 1 88kDa; beta catenin; Betacatenin; beta-catenin; beta-catenin 1; Bfc; C20orf33; Cadherin associated protein; catenin; catenin (cadherin associated protein), beta 1; catenin (cadherin associated protein), beta 1, 88kDa; catenin (cadherin-associated protein) beta 1; catenin (cadherin-associated protein), beta 1; catenin (cadherin-associated protein), beta 1, 88kDa; Catenin b 1; Catenin b1; catenin beta; catenin beta 1; catenin beta 1 L homeolog; Catenin beta1; Catenin beta-1; Catenin β 1; Catenin β1; Catnb; CHBCAT; CTNB1; CTNNB; ctnnb1; ctnnb1.L; ctnnb1-a; ctnnb1-b; dJ633O20.1; DKFZp686D02253; FLJ25606; FLJ37923; id:ibd2058; Mesc; MRD19; NAP; NYD-SP19; OK/SW-cl.35; OTTHUMP00000209289; P14L; PP8304; PRO2286; RP5-1118M15.1; wu:fb73e10; wu:fi81c06; wu:fk25h01; XELAEV_18031149mg; β catenin; βcatenin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12387, 1499, 397657, 477032 | |
-20°C | |
Liquid |