missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Calcium Sensing Receptor (Thr888) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538432
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
Senses changes in the extracellular concentration of calcium ions. The activity of this receptor is mediated by a G- protein that activates a phosphatidylinositol-calcium second messenger system.Spezifikation
Phospho-Calcium Sensing Receptor (Thr888) | |
Polyclonal | |
Unconjugated | |
CASR | |
CASR, CaSR, Extracellular calcium-sensing receptor precursor, GPRC2A, PCAR1, Parathyroid Cell calcium-sensing receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12374, 24247, 846 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P41180, Q9QY96, P48442 | |
CASR | |
A synthetic phosphopeptide derived from human Calcium Sensing Receptor around the phosphorylation site of Thr888 (R-A-TP-L-R) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |