missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-EIF2S1 (Ser52) Monoclonal Antibody (S.674.5), Invitrogen™
Rabbit Monoclonal Antibody
Marke: Invitrogen MA515133
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
Phospho-EIF2S1 (Ser52) Monoclonal antibody specifically detects Phospho-EIF2S1 (Ser52) in Drosophila, Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
Phospho-EIF2S1 (Ser52) | |
Monoclonal | |
68 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P05198, P68101, Q6ZWX6 | |
EIF2S1 | |
Synthetic phosphopeptide corresponding to residues surrounding pSer52 of human eIF2alpha | |
100 μL | |
Primary | |
Drosophila, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
S.674.5 | |
Unconjugated | |
EIF2S1 | |
0910001O23Rik; 2410026C18Rik; 35kDa; EIF2; EIF-2; eIF2 alpha; eIF2 alpha subunit; EIF2A; eIF-2A; eIF2alpha; EIF-2alpha; eIF-2-alpha; EIF2S1; eif2s1b; Eukaryotic translation initiation factor 2 subunit 1; eukaryotic translation initiation factor 2 subunit 1b; eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha; eukaryotic translation initiation factor 2, subunit 1 alpha b; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; fb19d01; translation initiation factor subunit; wu:fb19d01 | |
Rabbit | |
Affinity chromatography | |
RUO | |
13665, 1965, 54318 | |
-20°C | |
Liquid |