Learn More
Phospho-EIF2S1 (Ser51) Recombinant Rabbit Monoclonal Antibody (10H21L20), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen 701268
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with mouse, rat, non-human primate, rabbit and equine based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Phospho-EIF2S1 (Ser51) Recombinant Monoclonal antibody specifically detects Phospho-EIF2S1 (Ser51) in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Phospho-EIF2S1 (Ser51) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
0910001O23Rik; 2410026C18Rik; 35kDa; EIF2; EIF-2; eIF2 alpha; eIF2 alpha subunit; EIF2A; eIF-2A; eIF2alpha; EIF-2alpha; eIF-2-alpha; EIF2S1; eif2s1b; Eukaryotic translation initiation factor 2 subunit 1; eukaryotic translation initiation factor 2 subunit 1b; eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha; eukaryotic translation initiation factor 2, subunit 1 alpha b; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; fb19d01; translation initiation factor subunit; wu:fb19d01 | |
Phosphopeptide corresponding to amino acids 48–57 of human Eukaryotic translation initiation factor 2A [pS51] | |
100 μg | |
Primary | |
Human | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
10H21L20 | |
Unconjugated | |
P05198 | |
Rabbit | |
Protein A | |
RUO | |
1965 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |