Learn More
Phospho-ELK1 (Ser383) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA14625
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine, mouse, non-human primate and zebrafish based on 100% sequence homology. This antibody is specific for ∽46 kDa Elk-1 phosphorylated at Ser383 in Western blots using phospho and dephospho Elk-1 recombinant protein. Immunolabeling is blocked by the phosphopeptide used as the antigen but not by the corresponding dephosphopeptide.
Phospho-ELK1 (Ser383) Polyclonal antibody specifically detects Phospho-ELK1 (Ser383) in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
Phospho-ELK1 (Ser383) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
A4GTP4, P19419, P41969 | |
ELK1 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser383 of human ELK1 conjugated to KLH | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ELK1 | |
ELK 1; ELK1; Elk-1; ELK1, ETS transcription factor; ELK1, member of ETS oncogene family; ETS domain-containing protein Elk-1; ETS transcription factor ELK1; ETS-like gene 1; Oncogene Elk1; tyrosine kinase (ELK1) oncogene | |
Rabbit | |
Affinity chromatography | |
RUO | |
13712, 2002, 314436 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |