Learn More
Phospho-GSK3B (Ser9) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA14688
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with human, mouse, chicken, bovine, canine, non-human primate, zebrafish and Xenopus based on 100% sequence homology. In Western blot, this antibody is specific for the ∽46kDa GSK3 beta protein phosphorylated at Ser9. It also weakly labels the ∽51 kDa GSK3a band due to the high degree of homology between the 2 subunits. Immunolabeling is blocked by the phosphopeptide used as antigen but not by the corresponding dephosphopeptide.
Phospho-GSK3B (Ser9) Polyclonal antibody specifically detects Phospho-GSK3B (Ser9) in Mouse, Rat samples. It is validated for Western Blot.Spezifikation
Phospho-GSK3B (Ser9) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P18266, Q9WV60 | |
GSK3B | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser9 of rat GSK3B conjugated to KLH | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GSK3B | |
7330414F15Rik; 8430431H08Rik; C86142; EC 2.7.11.26; FA; Factor A; fb68h05; fk80d11; glycogen synthase kinase 3; glycogen synthase kinase 3 beta; glycogen synthase kinase 3 beta L homeolog; glycogen synthase kinase 3 beta variant 1; glycogen synthase kinase 3 beta variant 2; glycogen synthase kinase 3 beta variant 3; glycogen synthase kinase 3 beta variant 4; glycogen synthase kinase 3 beta variant 5; glycogen synthase kinase 3 beta, genome duplicate a; glycogen synthase kinase-3 beta; glycogen synthase kinase-3 beta splice variant X1; glycogen synthase kinase-3 beta splice variant X2; glycogen synthase kinase-3 beta splice variant X3; GSK 3 beta; GSK 3B; gsk3; GSK-3; GSK3 beta; GSK-3 beta; GSK-3 beta GSK3B; GSK-3[b]; Gsk3b; GSK-3b; gsk3b.L; gsk3ba; gsk3beta; GSK-3beta; gsk3-beta; GSK3beta isoform; intracellular kinase; pp37; serine/threonine-protein kinase GSK3B; wu:fb68h05; wu:fk80d11; XELAEV_180112401mg; xgsk-3; Xgsk-3 protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
56637, 84027 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |