Learn More
Phospho-HIRA (Thr555) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538579
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
HIRA is a histone chaperone that preferentially places the variant histone H3.3 in nucleosomes. Orthologs of this protein in yeast, flies, and plants are necessary for the formation of transcriptionally silent heterochomatin. It plays an important role in the formation of the senescence-associated heterochromatin foci. These foci likely mediate the irreversible cell cycle changes that occur in senescent cells. It is considered the primary candidate protein in some haploinsufficiency syndromes such as DiGeorge syndrome, and insufficient production of the gene may disrupt normal embryonic development.Spezifikation
Phospho-HIRA (Thr555) | |
Polyclonal | |
Unconjugated | |
HIRA | |
TUP1-like enhancer of split protein 1, DGCR1, HIR, TUPLE1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15260, 7290 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P54198, Q61666 | |
HIRA | |
A synthetic phosphopeptide derived from human HIRA around the phosphorylation site of Thr555 (L-T-T P-P-S) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |