missing translation for 'onlineSavingsMsg'
Learn More

H3K9acS10ph Monoclonal Antibody (APH3-64), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA125461

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11534942

  • 473.00 € / 50 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: Jurkat treated with Nocodazole. MA1-25461 is expected to cross react with mouse, rat, Xenopus laevis, bovine, C. elegans, chicken, Drosophilia melanogaster, and a wide range of other species due to sequence homolog Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

H3K9acS10ph Monoclonal antibody specifically detects H3K9acS10ph in Amphibian, Bovine, C. elegans, Chicken, Drosophila, Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

H3K9acS10ph
Monoclonal
2 mg/mL
0.01M PBS with 15mM sodium azide; pH 7.4
P09588, P68431, P68433, P84228, Q71DI3
H3C1, H3C10, H3C12, H3C14, H3C15, H3C2, H3C7, his-3, His3:CG31613, LOC107131750
Synthetic, acetylated and phosphorylated (acetyl K9) & (phospho S10) peptide, conjugated to KLH, corresponding to the N-terminal residues 7-20 of huma
50 μL
Primary
Amphibian, Bovine, C. elegans, Chicken, Drosophila, Human, Mouse, Rat
Antibody
IgG2a
ELISA, Immunoprecipitation, Western Blot
APH3-64
Unconjugated
H3C1
Acetyl H3; Acetyl Histone H3; B0035.7; CELE_T10C6.12; CG31613; CG31613-PA; CG33803; CG33806; CG33809; CG33812; CG33815; CG33818; CG33821; CG33824; CG33827; CG33830; CG33833; CG33836; CG33839; CG33842; CG33845; CG33848; CG33851; CG33854; CG33857; CG33860; CG33863; CG33866; Dmel\CG31613; Dmel_CG31613; F07B7.10; F07B7.3; F08G2.2; F17E9.13; F35H10.1; F45F2.4; F54E12.5; F55G1.10; H02I12.7; h3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3C; H3 K9; H3 K9ac; H3 S10; H3 S10ph; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; h3.2a; H3.3 histone A; H3.3 histone B; H3.3A; H3.3b; H3.5; H3.5 histone; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-143; H3-291; H3-3A; H3-3B; H3-5; H3-53; H3-614; H3a; H3b; H3-B; H3C1; H3c10; H3
Mouse
Antigen affinity chromatography
RUO
107131750, 126961, 180073, 260423, 318847, 319150, 319152, 333932, 360198, 616800, 788250, 8350, 8356, 8357, 8358, 8968, 97114
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote