missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Histone H3 (Thr11) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517360
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other phosphorylated histones or with acetylated histones.
Phospho-Histone H3 (Thr11) Polyclonal antibody specifically detects Phospho-Histone H3 (Thr11) in Human, Mouse, Rat, X. laevis samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
Phospho-Histone H3 (Thr11) | |
Polyclonal | |
Unconjugated | |
H3C1 | |
BUR5; CG31613; CG31613-PA; CG33803; CG33806; CG33809; CG33812; CG33815; CG33818; CG33821; CG33824; CG33827; CG33830; CG33833; CG33836; CG33839; CG33842; CG33845; CG33848; CG33851; CG33854; CG33857; CG33860; CG33863; CG33866; CG5825; CG5825-PA; CG5825-PC; CG5825-PD; CG8989; dH3.3A; Dmel\CG31613; Dmel\CG5825; Dmel_CG31613; Dmel_CG5825; fb58e10; h3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3B.1; H3 histone, family 3C; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; h3.2a; H3.3; H3.3 histone A; H3.3 histone B; H3.3a; H3.3B; H3.5; H3.5 histone; H3.A; H3.B; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-143; H3-291; H3-3A; H3-3B; H3-5; H3-53; H3-614; H3a; H3b; H3-B; H3C1; H3c10; H3c11; H3C12; H3C2; H3C3; H3C4; H3C6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126961, 260423, 319150, 319152, 333932, 360198, 399088, 679994, 8350, 8356, 8357, 8358, 8968, 97114 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
21 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P68431, P68433, P84228, Q71DI3 | |
H3C1, H3C10, H3C12, H3C14, H3C15, H3C2, H3C7, h3c8.S, HIST1H3A | |
Synthetic phosphopeptide corresponding to residues surrounding pThr11 of human histone H3 | |
100 μL | |
Primary | |
Human, Mouse, Rat, X. laevis | |
Antibody | |
IgG |