missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-HSL (Ser563) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517488
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with Ser565 phosphorylated HSL.
Phospho-HSL (Ser563) Polyclonal antibody specifically detects Phospho-HSL (Ser563) in Mouse samples. It is validated for Western Blot.Spezifikation
Phospho-HSL (Ser563) | |
Polyclonal | |
Unconjugated | |
LIPE | |
4933403G17Rik; AOMS4; EC 3.1.1.79; FPLD6; hormone - sensitive lipase testicular isoform; hormone-sensitive lipase; hormone-sensitive lipase testicular isoform; HSL; HSL antibody; LHS; lipase E; lipase E, hormone sensitive type; lipase, hormone sensitive; lipase, hormone-sensitive; LIPE; LIPS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16890 | |
-20°C | |
Liquid |
Western Blot | |
41 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P54310 | |
LIPE | |
Synthetic phosphopeptide corresponding to residues surrounding pSer552 of human HSL (equivalent to pSer563 of rat HSL) | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |