Learn More
Phospho-HSP27 (Ser15) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA1016
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-016 detects phosphorylated heat shock protein 27 (hsp27) from rat and human tissue tissues. PA1-016 has been successfully used in Western blot, immunoprecipitation, and immunofluorescence procedures. By Western blot, this antibody detects a ∽27 kDa protein which corresponds to phospho-hsp27 (Ser15) from HeLa cells. For immunolocalization, Cat. # PA1-018 is recommended. The PA1-016 immunogen is a synthetic phosphopeptide corresponding to residues L(10) L R G P (pS) W D P F R C(21) of human HSP27. The immunizing peptide is 81% conserved in mice, rats, and hamsters. The phosphorylated peptide (Cat. # PEP-187) and non-phosphorylated peptide (Cat. # PEP-240) are available for use in neutralization and control experiments.
Phospho-HSP27 (Ser15) Polyclonal antibody specifically detects Phospho-HSP27 (Ser15) in Human, Rat samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Spezifikation
Phospho-HSP27 (Ser15) | |
Polyclonal | |
Unconjugated | |
HSPB1 | |
27kDa; 28 kDa heat shock protein; CMT2F; DKFZp586P1322; epididymis secretory protein Li 102; estrogen-regulated 24 kDa protein; Growth-related 25 kDa protein; heat shock 25 kDa protein; Heat shock 27 kDa protein; heat shock 27kD protein 1; heat shock 27kDa protein 1; Heat shock protein; heat shock protein 1; heat shock protein 27 kDa beta-1; heat shock protein B1; heat shock protein beta-1; heat shock protein family B (small) member 1; heat shock protein, 25 kDa; heat-shock protein; HEL-S-102; HMN2B; HS; HS.76067; HSP; HSP 25; HSP 27; Hsp25; HSP27; HSP28; HSPB1; p25; Phospho-HSP 27; SRP27; Stress-responsive protein 27; truncated hsp25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
24471, 3315 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P04792, P42930 | |
HSPB1 | |
Synthetic phosphopeptide corresponding to residues L(10) L R G P (pS) W D P F R C(21) of human HSP27. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |