missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-IkB alpha (Ser32) Monoclonal Antibody (B.142.7), Invitrogen™
Rabbit Monoclonal Antibody
Marke: Invitrogen MA515087
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
Phospho-IkB alpha (Ser32) Monoclonal antibody specifically detects Phospho-IkB alpha (Ser32) in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunoprecipitation, Western Blot.Spezifikation
Phospho-IkB alpha (Ser32) | |
Monoclonal | |
218.3 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P25963, Q63746, Q9Z1E3 | |
NFKBIA | |
Synthetic phosphopeptide corresponding to residues surrounding pSer32 of human IkB alpha. | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
B.142.7 | |
Unconjugated | |
NFKBIA | |
AI462015; ECI-6; ECI-6/IKBA; I kappa B-alpha; I(Kappa)B(alpha); I79_018146; IkappaB alpha; IkappaBalpha; I-kappaBalpha; I-kappa-B-alpha; Ikba; IKBalpha; IKB-alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells, alpha; MAD3; MAD-3; Major histocompatibility complex enhancer-binding protein MAD3; NF kappa B inhibitor alpha; NF-kappaB inhibitor alpha; NF-kappa-B inhibitor alpha; NFKB inhibitor alpha; NFKBI; Nfkbia; nuclear factor of kappa light chain gene enhancer in B-cells; nuclear factor of kappa light chain gene enhancer in B-cells inhibitor, alpha; nuclear factor of kappa light polyp gene enhancer in B-cell 1; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, alpha; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha; Rel-associated pp40; REL-associated protein pp40; RL/IF-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
18035, 25493, 4792 | |
-20°C | |
Liquid |