missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-IKK alpha/beta (Ser180, Ser181) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538255
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
The NF-kB/Rel transcription factors are present in the cytosol in an inactive state, complexed with the inhibitory IkB proteins. Most agents that activate NF-kB do so through a common pathway based on phosphorylation-induced, proteasome-mediated degradation of IkB. The key regulatory step in this pathway involves activation of a high molecular weight IkB kinase (IKK) complex whose catalysis is generally carried out by three tightly associated IKK subunits. IKK alpha and IKK beta serve as the catalytic subunits of the kinase and IKK gamma serves as the regulatory subunit. Activation of IKK depends upon phosphorylation at Ser177 and Ser181 in the activation loop of IKK beta (Ser176 and Ser180 in IKK alpha), which causes conformational changes, resulting in kinase activation.Spezifikation
Phospho-IKK alpha/beta (Ser180, Ser181) | |
Polyclonal | |
Unconjugated | |
CHUK | |
I kappa-B kinase alpha, IkBKA, IKK-alpha, IKK-A, IkappaB kinase, EC 2.7.11.10, I-kappa-B kinase 1, IKK1, Conserved helix-loop-helix ubiquitous kinase, Nuclear factor NF-kappa-B inhibitor kinase alpha, NFKBIKA, CHUK, IKKA, TCF16, IKK-beta, IKK2, IKKB, NFKBIKB; I-kappa-B kinase 2; I-kappa-B-kinase beta; IKK-B; inhibitor of nuclear factor kappa-B kinase subunit beta; nuclear factor NF-kappa-B inhibitor kinase beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1147, 12675, 16150, 3551 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O14920, O15111, O88351, Q60680 | |
CHUK, IKBKB | |
A synthetic phosphopeptide derived from human IKK-alpha/beta around the phosphorylation site of Ser180/181 (C-T-SP-F-V) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |