Learn More
Phospho-INSR (Tyr1334) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen 44809G
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
This antibody has been negatively preadsorbed using a non-phosphopeptide corresponding to the site of phosphorylation to remove antibody that is reactive with non-phosphorylated Insulin Receptor. The final product is generated by affinity chromatography using a Insulin Receptor-derived peptide that is phosphorylated at tyrosine 1334. This antibody recognizes human insulin receptor phosphorylated at tyrosine 1334 as well as the human IR Isoform B phosphorylated at tyrosine 1332. Western blot positive control used was insulin-treated CHO-T cells expressing Human Insulin Receptor.
Phospho-INSR (Tyr1334) Polyclonal antibody specifically detects Phospho-INSR (Tyr1334) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
Phospho-INSR (Tyr1334) | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
P06213, P15127, P15208 | |
INSR | |
The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human Insulin Receptor that contains tyrosine 1334. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Unconjugated | |
INSR | |
4932439J01Rik; CD 220; CD220; CD220 antigen; CD220 beta; CD221; D630014A15Rik; HHF 5; HHF5; IGF-I receptor; INSR; Insulin receptor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR 1; IR beta; IR1; IR-1; IR-A; IR-B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16337, 24954, 3643 | |
-20°C | |
Liquid |