Learn More
Phospho-IP3 Receptor 1 (Ser1588, Ser1598) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538628
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
Phospho-IP3 Receptor 1 (Ser1588, Ser1598) Polyclonal antibody specifically detects Phospho-IP3 Receptor 1 (Ser1588, Ser1598) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Peptide ELISA.Spezifikation
Phospho-IP3 Receptor 1 (Ser1588, Ser1598) | |
Polyclonal | |
Unconjugated | |
Itpr1 | |
ACV; CLA4; D6Pas2; ENSMUSG00000072853; Gm10429; I145TR; inositol 1,4,5-triphosphate receptor 1; inositol 1,4,5-triphosphate receptor, type 1; inositol 1,4,5-trisphosphate receptor 1; inositol 1,4,5-trisphosphate receptor type 1; inositol 1,4,5-trisphosphate receptor, type 1; inositol 1,4,5-trisphosphate-binding protein P400; InsP>3<R type I; Insp3r; InsP3R type I; insP3R1; IP3 receptor; IP3 receptor isoform 1; Ip3r; IP-3-R; IP3R 1; IP3R1; Itpr1; Itpr-1; opisthotonus; opt; P400; Pcd6; Pcp1; Pcp-1; PPP1R94; protein PCD-6; protein phosphatase 1, regulatory subunit 94; Purkinje cell protein 1; SCA15; SCA16; SCA29; SI-SIII-SIIA; Type 1 inositol 1,4,5-trisphosphate receptor; type 1 InsP3 receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16438, 25262, 3708 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Peptide ELISA | |
1 mg/mL | |
Dulbecco's PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P11881, P29994, Q14643 | |
Itpr1 | |
A synthetic phosphopeptide derived from human Inositol 1,4,5-trisphosphate R1 around the phosphorylation site of Ser1598/1588 (R-D-SP-V-L) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |