missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-IRS1 (Ser794) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538364
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
IRS-1, a major substrate of the insulin receptor, is phosphorylated in response to stimulation of cells by insulin, insulin-like growth factor 1 (IGF-1) and interleukin 4 (IL-4). IRS-1 is phosphorylated on serine, threonine and tyrosine residues in a variety of tissues. An insulin-sensitive serine/threonine kinase casein kinase II mediates a portion of the insulin-stimulated serine/threonine phosphorylation of overexpressed IRS-1 in vivo. Thr 502 is identified as the major casein kinase II-catalyzed phosphorylation site in rat IRS-1, and Ser 99 is an additional phosphorylation site catalyzed by casein kinase II. Thus, casein kinase II-catalyzed phosphorylation of IRS-1 may be a component of the intracellular insulin signaling cascade. IRS-1 contains three putative binding sites for 14-3-3 (Ser 270, Ser 374 and Ser 641) and the motif around Ser 270 is located in the phosphortyrosine binding domain of IRS-1, which is responsible for the interaction with the insulin receptor.Spezifikation
Phospho-IRS1 (Ser794) | |
Polyclonal | |
Unconjugated | |
IRS1 | |
Insulin receptor substrate 1, IRS-1, IRS1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16367, 25467, 3667 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P35568, P35569, P35570 | |
IRS1 | |
A synthetic phosphopeptide derived from human IRS-1 around the phosphorylation site of Ser794 (S-T-SP-S-G) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |