Learn More
Phospho-IRS1 (Ser616) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen 44550G
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
Purified from rabbit serum by sequential epitope-specific chromatography, this product contains enough material for 10 mini-blots. This antibody has been negatively preadsorbed using a non-phosphopeptide corresponding to the site of phosphorylation to remove antibody that is reactive with non-phosphorylated insulin receptor substrate-1 (IRS-1). The final product is generated by affinity chromatography using IRS-1 derived peptide that is phosphorylated at serine 616. The antibody has been used in western blotting. Previous lots of this antibody have been used in immunohistochemistry. In western analysis the positive control used was human embryonic kidney cell line transfected with a vector encoding IRS-1 (293T) and stimulated with a phorbol ester, TPA.
Phospho-IRS1 (Ser616) Polyclonal antibody specifically detects Phospho-IRS1 (Ser616) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Phospho-IRS1 (Ser616) | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide; pH 7.3 | |
P35568, P35569, P35570 | |
IRS1 | |
The antiserum was produced against a chemically synthesized phosphopeptide derived from a region of IRS-1 that contains serine 616. The sequence is conserved in many species including human, mouse, and rat | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
IRS1 | |
G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16367, 25467, 3667 | |
-20°C | |
Liquid |